Assay Details
| Gene Symbol: Il20ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172786.2 | NP_766374.1 | 5 - 6 | 780 | 108 | |
| GenBank mRNA | AK054215.1 | - | 5 - 6 | 781 | 108 | |
| BC056628.1 | - | 5 - 6 | 764 | 5033177 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il20ra
Genomic Map