Assay Details
| Gene Symbol: Slc16a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114332.1 | NP_001107804.1 | 3 - 4 | 1153 | 99 | |
| GenBank mRNA | AK036743.1 | - | 3 - 4 | 1154 | 99 | |
| AK144135.1 | - | 3 - 4 | 1156 | 99 | ||
| AK151290.1 | - | 3 - 4 | 1154 | 99 | ||
| AK151636.1 | - | 3 - 4 | 1155 | 99 | ||
| AK151704.1 | - | 3 - 4 | 1158 | 99 | ||
| AK165240.1 | - | 3 - 4 | 1154 | 99 | ||
| BC052877.1 | - | 3 - 4 | 634 | 30046388 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc16a10
Genomic Map