Assay Details
| Gene Symbol: Slc16a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114332.1 | NP_001107804.1 | 5 - 6 | 1526 | 118 | |
| GenBank mRNA | AK036743.1 | - | 5 - 6 | 1527 | 118 | |
| AK144135.1 | - | 5 - 6 | 1531 | 120 | ||
| AK151704.1 | - | 5 - 6 | 1531 | 118 | ||
| AK165240.1 | - | 5 - 6 | 1527 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc16a10
Genomic Map