Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400029063.mm.1 |
TC0400003573.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029063.mm.1 |
NM_029137.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029063.mm.1 |
NM_029137.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029061.mm.1 |
TC0400003573.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029061.mm.1 |
NM_029137.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029061.mm.1 |
NM_029137.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029062.mm.1 |
TC0400003573.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029062.mm.1 |
NM_029137.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029062.mm.1 |
NM_029137.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029065.mm.1 |
TC0400003573.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029065.mm.1 |
NM_029137.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029065.mm.1 |
NM_029137.1 |
Refseq |