Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1432537_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432537_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432537_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
74989_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
74989_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0400029060.mm.1 |
TC0400003573.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029060.mm.1 |
NM_029137.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029060.mm.1 |
NM_029137.1 |
Refseq |
Mouse Expression Set 430 |
1432537_at |
- |
- |
Mouse Expression Set 430 |
1432537_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1432537_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1432537_PM_at |
- |
- |