Assay Details
| Gene Symbol: Gprc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153071.1 | NP_694711.1 | 5 - 6 | 1702 | 71 | |
| GenBank mRNA | AY101365.1 | - | 5 - 6 | 1702 | 71 | |
| AY735399.1 | - | 5 - 6 | 1675 | 71 | ||
| AY735400.1 | - | 5 - 6 | 1150 | 71 | ||
| AY735401.1 | - | 4 - 5 | 1462 | 71 | ||
| BC108998.2 | - | 5 - 6 | 1702 | 40048019 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097645 | AY101365.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097645 | NM_153071.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324837 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346791 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921732 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131042 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4984 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73636 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29982 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34521 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117536 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136191 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160926 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097645 | AY101365.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097645 | NM_153071.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P339038 | BC108998.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P339038 | AY735400.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P339038 | AY735401.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P339038 | AY101365.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P339038 | AY735399.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27134074 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02123704 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160926 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136191 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117536 | - | - |
| Whole Mouse Genome G4122A | A_51_P339038 | BC108998.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P339038 | AY735400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P339038 | AY735401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P339038 | AY101365.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P339038 | AY735399.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130258 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117536 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136191 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160926 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08760617 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08760619 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08760622 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08760626 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08760618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27134026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27133994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27133992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27133993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27133996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27133994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27133999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08760626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08516557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08272490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08616493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08856744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08355561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27134058 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10369029 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369030 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369037 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369035 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369031 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369032 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369033 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369034 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369036 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428053_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10369028 | - | - |
| Mouse Genome 430 2.0 Array | 1428053_at | - | - |
| Mouse Genome 430 2.0 Array | 1428053_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210198_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210198_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000015055.mm.1 | NM_153071.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000015055.mm.1 | NM_153071.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000015055.mm.1 | TC1000002148.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428053_PM_at | - | - |
| Mouse Expression Set 430 | 1428053_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428053_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428053_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428053_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428053_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4560576 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4560576 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004560576 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2588955 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2588955 | - | - |
| mouse-6 v1.1 (Illumina) | scl37976.6.1_275-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4560576 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37976.6.1_275-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4560576 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2588955 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2588955 | - | - |

Gprc6a
Genomic Map