| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000015314.mm.1 |
NM_001033385.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000015314.mm.1 |
NM_001033385.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000015314.mm.1 |
TC1000002185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000015316.mm.1 |
NM_001033385.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000015316.mm.1 |
NM_001033385.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000015316.mm.1 |
TC1000002185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000015313.mm.1 |
NM_001033385.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000015313.mm.1 |
NM_001033385.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000015313.mm.1 |
TC1000002185.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000015315.mm.1 |
NM_001033385.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000015315.mm.1 |
NM_001033385.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000015315.mm.1 |
TC1000002185.mm.1 |
MTA TC |