Assay Details
| Gene Symbol: Tbc1d32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033385.3 | NP_001028557.2 | 17 - 18 | 2069 | 100 | |
| GenBank mRNA | AK141206.1 | - | 2 - 3 | 264 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d32
Genomic Map