Assay Details
| Gene Symbol: Aunip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081099.1 | NP_001074568.1 | 2 - 3 | 301 | 109 | |
| GenBank mRNA | AK011279.1 | - | 2 - 3 | 226 | 109 | |
| BC138271.1 | - | 2 - 3 | 277 | 8861290 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56397 | BC138271.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56397 | NM_001081099.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56397 | AK011279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56397 | BC138271.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56397 | NM_001081099.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56397 | AK011279.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278413 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386948 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71511 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56397 | BC138271.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56397 | NM_001081099.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56397 | AK011279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56397 | BC138271.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56397 | NM_001081099.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56397 | AK011279.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00994907 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05412117 | - | - |
| Whole Mouse Genome G4122A | A_52_P56397 | BC138271.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56397 | NM_001081099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56397 | AK011279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56397 | BC138271.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56397 | NM_001081099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56397 | AK011279.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00994907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03689896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03627454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03689894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03689888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03354402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03689886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03627452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323196 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10508999 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508997 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508998 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429812_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10508996 | - | - |
| Mouse Genome 430 2.0 Array | 1458905_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1429812_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445293_at | - | - |
| Mouse Genome 430 2.0 Array | 1445293_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429812_at | - | - |
| Mouse Genome 430 2.0 Array | 1458905_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69885_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69885_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400013079.mm.1 | NM_001081099.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400013079.mm.1 | NM_001081099.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400013079.mm.1 | TC0400001642.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400013081.mm.1 | NM_001081099.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400013081.mm.1 | NM_001081099.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400013081.mm.1 | TC0400001642.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400013080.mm.1 | NM_001081099.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400013080.mm.1 | NM_001081099.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400013080.mm.1 | TC0400001642.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400013082.mm.1 | NM_001081099.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400013082.mm.1 | NM_001081099.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400013082.mm.1 | TC0400001642.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429812_at | - | - |
| Mouse Expression Set 430 | 1429812_PM_at | - | - |
| Murine Genome U74v2 Set | 169224_f_at | - | - |
| Murine Genome U74v2 Set | 171618_PM_f_at | - | - |
| Murine Genome U74v2 Set | 169224_PM_f_at | - | - |
| Murine Genome U74v2 Set | 171618_f_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429812_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429812_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 308384 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2588581 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2588581 | - | - |

Aunip
Genomic Map