Assay Details
| Gene Symbol: Arhgap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176837.2 | NP_789807.1 | 6 - 7 | 1022 | 111 | |
| GenBank mRNA | AK014728.1 | - | 6 - 7 | 970 | 111 | |
| AK078522.1 | - | 1 - 2 | 74 | 111 | ||
| AK087800.1 | - | 6 - 7 | 998 | 111 | ||
| AK162202.1 | - | 2 - 3 | 265 | 111 | ||
| BC030858.1 | - | 6 - 7 | 1022 | 4194996 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap18
Genomic Map