Assay Details
| Gene Symbol: Il9r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134458.1 | NP_001127930.1 | 3 - 4 | 310 | 79 | |
| NM_008374.2 | NP_032400.1 | 3 - 4 | 310 | 79 | ||
| GenBank mRNA | M84746.1 | - | 3 - 4 | 253 | 79 | |
More Information
Set Membership: |
|

Il9r
Genomic Map