Assay Details
| Gene Symbol: Il9r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008374.2 | NP_032400.1 | 5 - 6 | 640 | 76 | |
| GenBank mRNA | M84746.1 | - | 5 - 6 | 583 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il9r
Genomic Map