Assay Details
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290676.1 | NP_001277605.1 | 3 - 4 | 2607 | 83 | |
GenBank mRNA | BC079599.1 | - | 3 - 4 | 415 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
