Assay Details
| Gene Symbol: Rhbdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291818.1 | NP_001278747.1 | 13 - 14 | 1754 | 55 | |
| NM_010117.2 | NP_034247.2 | 13 - 14 | 1873 | 55 | ||
| GenBank mRNA | AK144437.1 | - | 13 - 14 | 1742 | 55 | |
| BC023469.1 | - | 8 - 9 | 1021 | 55 | ||
| BC027346.1 | - | 13 - 14 | 1818 | 4011421 | 55 | |
| M99623.1 | - | 3 - 4 | 194 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the M99623.1 transcripts.
Set Membership: |
|

Rhbdf1
Genomic Map