Assay Details
| Gene Symbol: Rhbdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291818.1 | NP_001278747.1 | 7 - 8 | 985 | 72 | |
| NM_010117.2 | NP_034247.2 | 7 - 8 | 1104 | 72 | ||
| GenBank mRNA | AK144437.1 | - | 7 - 8 | 973 | 72 | |
| BC023469.1 | - | 2 - 3 | 252 | 72 | ||
| BC027346.1 | - | 7 - 8 | 1049 | 4011421 | 72 | |
| CD554689.1 | - | 3 - 4 | 402 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rhbdf1
Genomic Map