Assay Details
| Gene Symbol: Fbxw11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271347.1 | NP_001258276.1 | 9 - 10 | 1609 | 73 | |
| NM_001271348.1 | NP_001258277.1 | 9 - 10 | 1570 | 73 | ||
| NM_001271349.1 | NP_001258278.1 | 8 - 9 | 1507 | 73 | ||
| NM_134015.3 | NP_598776.1 | 10 - 11 | 1672 | 73 | ||
| GenBank mRNA | AB093260.1 | - | 8 - 9 | 1314 | 73 | |
| AK041532.1 | - | 9 - 10 | 1402 | 73 | ||
| AK149139.1 | - | 9 - 10 | 1392 | 73 | ||
| AK152181.1 | - | 9 - 10 | 1332 | 73 | ||
| AK166226.1 | - | 9 - 10 | 1267 | 73 | ||
| AK168667.1 | - | 8 - 9 | 1324 | 73 | ||
| AY038079.1 | - | 10 - 11 | 1336 | 73 | ||
| BC008552.1 | - | 3 - 4 | 405 | 73 | ||
| BC034261.1 | - | 10 - 11 | 1352 | 4237375 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxw11
Genomic Map