Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00004464.mm.1 |
NM_007435.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004464.mm.1 |
TC0X00000691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004463.mm.1 |
NM_007435.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004463.mm.1 |
TC0X00000691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004466.mm.1 |
NM_007435.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004466.mm.1 |
TC0X00000691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004468.mm.1 |
NM_007435.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004468.mm.1 |
TC0X00000691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004462.mm.1 |
NM_007435.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004462.mm.1 |
TC0X00000691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004469.mm.1 |
NM_007435.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004469.mm.1 |
TC0X00000691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004465.mm.1 |
NM_007435.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004465.mm.1 |
TC0X00000691.mm.1 |
MTA TC |