Assay Details
Gene Symbol: Opn1mw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008106.2 | NP_032132.1 | 3 - 4 | 608 | 64 | |
GenBank mRNA | AF011389.1 | - | 3 - 4 | 594 | 64 | |
AF190672.1 | - | 3 - 4 | 610 | 64 | ||
AK149287.1 | - | 3 - 4 | 609 | 64 | ||
BC014826.1 | - | 3 - 4 | 588 | 4507311 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
