Assay Details
| Gene Symbol: Hdac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008229.2 | NP_032255.2 | 11 - 12 | 1431 | 60 | |
| GenBank mRNA | AB221663.1 | - | 11 - 12 | 1223 | 60 | |
| AK135572.1 | - | 6 - 7 | 737 | 60 | ||
| AK141649.1 | - | 1 - 2 | 48 | 60 | ||
| BC138517.1 | - | 11 - 12 | 1403 | 8861537 | 60 | |
| U31758.1 | - | 11 - 12 | 1391 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hdac2
Genomic Map