Assay Details
| Gene Symbol: Hdac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008229.2 | NP_032255.2 | 3 - 4 | 495 | 149 | |
| GenBank mRNA | AB221663.1 | - | 3 - 4 | 287 | 149 | |
| AK051753.1 | - | 3 - 4 | 470 | 149 | ||
| BC138517.1 | - | 3 - 4 | 467 | 8861537 | 149 | |
| U31758.1 | - | 3 - 4 | 455 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hdac2
Genomic Map