Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000014434.mm.1 |
NM_001113388.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014434.mm.1 |
TC1000002032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014433.mm.1 |
NM_001113388.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014433.mm.1 |
NM_001173500.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014433.mm.1 |
TC1000002032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014438.mm.1 |
NM_001001489.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014438.mm.1 |
NM_001001489.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014438.mm.1 |
NM_001113385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014438.mm.1 |
NM_001113385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014438.mm.1 |
NM_001113388.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014438.mm.1 |
TC1000002032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014432.mm.1 |
NM_001113388.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014432.mm.1 |
TC1000002032.mm.1 |
MTA TC |