Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400027152.mm.1 |
TC0400004219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027152.mm.1 |
NM_001253381.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027152.mm.1 |
NM_001253381.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027149.mm.1 |
TC0400004219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027149.mm.1 |
NM_001253381.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027149.mm.1 |
NM_001253381.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027149.mm.1 |
NM_017377.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027149.mm.1 |
NM_017377.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027149.mm.1 |
NR_045570.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027149.mm.1 |
NR_045570.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027147.mm.1 |
TC0400004219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027147.mm.1 |
NM_001253381.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027147.mm.1 |
NM_001253381.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027147.mm.1 |
NM_017377.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027147.mm.1 |
NM_017377.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027147.mm.1 |
NR_045570.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027147.mm.1 |
NR_045570.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027148.mm.1 |
TC0400004219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027148.mm.1 |
NM_001253381.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027148.mm.1 |
NM_001253381.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027148.mm.1 |
NM_017377.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027148.mm.1 |
NM_017377.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027148.mm.1 |
NR_045570.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027148.mm.1 |
NR_045570.1 |
Refseq |