Assay Details
| Gene Symbol: St3gal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161774.2 | NP_001155246.1 | 10 - 11 | 1127 | 77 | |
| NM_001285520.1 | NP_001272449.1 | 10 - 11 | 1457 | 77 | ||
| NM_001285521.1 | NP_001272450.1 | 9 - 10 | 1079 | 77 | ||
| NM_009176.5 | NP_033202.3 | 10 - 11 | 1471 | 77 | ||
| GenBank mRNA | AK005053.1 | - | 10 - 11 | 1416 | 77 | |
| AK013016.1 | - | 9 - 10 | 1018 | 77 | ||
| AK136442.1 | - | 10 - 11 | 1408 | 77 | ||
| BC006710.1 | - | 10 - 11 | 1095 | 3584323 | 77 | |
| X84234.1 | - | 10 - 11 | 950 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

St3gal3
Genomic Map