Assay Details
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007939.2 | NP_031965.2 | 15 - 16 | 2801 | 70 | |
GenBank mRNA | AK220393.1 | - | 13 - 14 | 2285 | 70 | |
U72207.1 | - | 15 - 16 | 2810 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
