Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900014272.mm.1 |
NM_016860.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014272.mm.1 |
NM_016860.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014272.mm.1 |
TC1900001603.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014273.mm.1 |
NM_016860.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014273.mm.1 |
NM_016860.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014273.mm.1 |
TC1900001603.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014274.mm.1 |
NM_016860.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014274.mm.1 |
NM_016860.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014274.mm.1 |
TC1900001603.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014275.mm.1 |
NM_016860.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014275.mm.1 |
NM_016860.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014275.mm.1 |
TC1900001603.mm.1 |
MTA TC |