Assay Details
| Gene Symbol: Actr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016860.1 | NP_058556.1 | 7 - 8 | 814 | 99 | |
| GenBank mRNA | AB010297.1 | - | 7 - 8 | 814 | 99 | |
| AK010632.1 | - | 7 - 8 | 811 | 99 | ||
| AK019518.1 | - | 7 - 8 | 818 | 99 | ||
| AK088062.1 | - | 7 - 8 | 821 | 99 | ||
| AK167450.1 | - | 7 - 8 | 821 | 99 | ||
| AK168889.1 | - | 7 - 8 | 818 | 99 | ||
| BC007131.1 | - | 7 - 8 | 829 | 3599656 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Actr1a
Genomic Map