Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029186.2 | NP_083462.2 | 2 - 3 | 344 | 88 | |
| GenBank mRNA | AK031554.1 | - | 2 - 3 | 290 | 88 | |
| AK151797.1 | - | 2 - 3 | 319 | 88 | ||
| AK161305.1 | - | 2 - 3 | 300 | 88 | ||
| AK170247.1 | - | 291 | 88 | |||
| BC031169.1 | - | 2 - 3 | 289 | 88 | ||
| BC058746.1 | - | 2 - 3 | 290 | 6399588 | 88 | |
More Information
Set Membership: |
|

Tmem180
Genomic Map