Assay Details
| Gene Symbol: Adcy8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009623.2 | NP_033753.2 | 11 - 12 | 3688 | 77 | |
| GenBank mRNA | BC059010.1 | - | 11 - 12 | 2785 | 77 | |
| BC094231.1 | - | 11 - 12 | 3126 | 77 | ||
| U85021.1 | - | 11 - 12 | 3548 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcy8
Genomic Map