Assay Details
Gene Symbol: Adcy8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291903.1 | NP_001278832.1 | 5 - 6 | 2668 | 64 | |
NM_009623.2 | NP_033753.2 | 5 - 6 | 2668 | 64 | ||
GenBank mRNA | AK138244.1 | - | 5 - 6 | 2669 | 64 | |
AK163443.1 | - | 5 - 6 | 2644 | 64 | ||
BC059010.1 | - | 5 - 6 | 1766 | 64 | ||
BC094231.1 | - | 5 - 6 | 2106 | 64 | ||
U85021.1 | - | 5 - 6 | 2528 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
