Assay Details
Gene Symbol: Slc30a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172816.3 | NP_766404.1 | 5 - 6 | 1060 | 140 | |
GenBank mRNA | AK050482.1 | - | 5 - 6 | 961 | 140 | |
AK050494.1 | - | 5 - 6 | 1060 | 140 | ||
BC117544.1 | - | 5 - 6 | 892 | 40105617 | 140 | |
BC118521.1 | - | 5 - 6 | 889 | 40105622 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
