Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500012531.mm.1 |
TC0500001411.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012531.mm.1 |
NM_026000.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012531.mm.1 |
NM_026000.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012527.mm.1 |
TC0500001411.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012527.mm.1 |
NM_026000.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012527.mm.1 |
NM_026000.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012532.mm.1 |
TC0500001411.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012532.mm.1 |
NM_026000.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012532.mm.1 |
NM_026000.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012524.mm.1 |
TC0500001411.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012524.mm.1 |
NM_026000.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012524.mm.1 |
NM_026000.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012526.mm.1 |
TC0500001411.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012526.mm.1 |
NM_026000.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012526.mm.1 |
NM_026000.2 |
Refseq |