Assay Details
| Gene Symbol: Rnf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030564.1 | NP_085041.1 | 4 - 5 | 840 | 154 | |
| GenBank mRNA | AF434815.1 | - | 3 - 4 | 735 | 154 | |
| AK045696.1 | - | 4 - 5 | 836 | 154 | ||
| AK046274.1 | - | 4 - 5 | 822 | 154 | ||
| AK046381.1 | - | 4 - 5 | 836 | 154 | ||
| AK077221.1 | - | 4 - 5 | 837 | 154 | ||
| AK132330.1 | - | 856 | 154 | |||
| AK137606.1 | - | 816 | 154 | |||
| AK150976.1 | - | 4 - 5 | 834 | 154 | ||
| AK159753.1 | - | 4 - 5 | 822 | 154 | ||
| BC004042.1 | - | 4 - 5 | 840 | 3496314 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf34
Genomic Map