Assay Details
| Gene Symbol: Bcl7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029850.3 | NP_084126.1 | 3 - 4 | 451 | 60 | |
| GenBank mRNA | AK014498.1 | - | 3 - 4 | 394 | 60 | |
| AK044782.1 | - | 3 - 4 | 422 | 60 | ||
| AK086803.1 | - | 3 - 4 | 448 | 60 | ||
| BC038481.1 | - | 3 - 4 | 376 | 4501205 | 60 | |
| BC046777.1 | - | 3 - 4 | 281 | 5709511 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcl7a
Genomic Map