Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500028798.mm.1 |
TC0500003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028798.mm.1 |
NM_001042491.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028798.mm.1 |
NM_001289517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028798.mm.1 |
NM_001289518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028798.mm.1 |
NM_001289519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028798.mm.1 |
NM_001289520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028798.mm.1 |
NM_021505.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028805.mm.1 |
TC0500003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028805.mm.1 |
NM_001042491.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028805.mm.1 |
NM_001289517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028805.mm.1 |
NM_001289518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028805.mm.1 |
NM_001289519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028805.mm.1 |
NM_001289520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028805.mm.1 |
NM_021505.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028797.mm.1 |
TC0500003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028797.mm.1 |
NM_001042491.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028797.mm.1 |
NM_001289517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028797.mm.1 |
NM_001289518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028797.mm.1 |
NM_001289519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028797.mm.1 |
NM_001289520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028797.mm.1 |
NM_021505.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028802.mm.1 |
TC0500003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028802.mm.1 |
NM_001042491.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028802.mm.1 |
NM_001289517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028802.mm.1 |
NM_001289518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028802.mm.1 |
NM_001289519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028802.mm.1 |
NM_001289520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028802.mm.1 |
NM_021505.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028804.mm.1 |
TC0500003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028804.mm.1 |
NM_001042491.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028804.mm.1 |
NM_001289517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028804.mm.1 |
NM_001289518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028804.mm.1 |
NM_001289519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028804.mm.1 |
NM_001289520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028804.mm.1 |
NM_021505.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028803.mm.1 |
TC0500003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028803.mm.1 |
NM_001042491.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028803.mm.1 |
NM_001289517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028803.mm.1 |
NM_001289518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028803.mm.1 |
NM_001289519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028803.mm.1 |
NM_001289520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028803.mm.1 |
NM_021505.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028807.mm.1 |
TC0500003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028807.mm.1 |
NM_001042491.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028807.mm.1 |
NM_001289517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028807.mm.1 |
NM_001289518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028807.mm.1 |
NM_001289519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028807.mm.1 |
NM_001289520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028807.mm.1 |
NM_021505.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028795.mm.1 |
TC0500003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028795.mm.1 |
NM_001042491.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028795.mm.1 |
NM_001289517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028795.mm.1 |
NM_001289518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028795.mm.1 |
NM_001289519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028795.mm.1 |
NM_001289520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028795.mm.1 |
NM_021505.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028800.mm.1 |
TC0500003220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500028800.mm.1 |
NM_001042491.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028800.mm.1 |
NM_001289517.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028800.mm.1 |
NM_001289518.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028800.mm.1 |
NM_001289519.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028800.mm.1 |
NM_001289520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500028800.mm.1 |
NM_021505.3 |
Refseq |