Assay Details
| Gene Symbol: Setd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040398.2 | NP_001035488.2 | 14 - 15 | 5799 | 74 | |
| GenBank mRNA | AK122435.1 | - | 6 - 7 | 2134 | 74 | |
| BC035291.1 | - | 6 - 7 | 2301 | 74 | ||
| BC038367.1 | - | 8 - 9 | 2877 | 74 | ||
| BC040775.1 | - | 6 - 7 | 2301 | 74 | ||
| BC041681.1 | - | 6 - 7 | 2320 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Setd1b
Genomic Map