Assay Details
Gene Symbol: Lrrc43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033461.3 | NP_001028633.1 | 10 - 11 | 1846 | 79 | |
NM_001289821.1 | NP_001276750.1 | 10 - 11 | 1825 | 79 | ||
GenBank mRNA | AK133051.1 | - | 10 - 11 | 1847 | 79 | |
AK167318.1 | - | 10 - 11 | 1824 | 79 | ||
AK167729.1 | - | 10 - 11 | 1824 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
