Assay Details
Gene Symbol: Lrrc43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033461.3 | NP_001028633.1 | 5 - 6 | 904 | 89 | |
NM_001289821.1 | NP_001276750.1 | 5 - 6 | 904 | 89 | ||
NM_001289822.1 | NP_001276751.1 | 5 - 6 | 904 | 89 | ||
GenBank mRNA | AK133051.1 | - | 5 - 6 | 905 | 89 | |
AK167318.1 | - | 5 - 6 | 903 | 89 | ||
AK167729.1 | - | 5 - 6 | 903 | 89 | ||
BC150919.1 | - | 5 - 6 | 918 | 9087830 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
