Assay Details
Gene Symbol: Kdm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013910.2 | NP_038938.1 | 6 - 7 | 1035 | 91 | |
GenBank mRNA | AK129479.1 | - | 17 - 18 | 2472 | 91 | |
BC057622.1 | - | 6 - 7 | 1030 | 6402440 | 91 | |
BC065090.1 | - | 17 - 18 | 2671 | 6851421 | 91 | |
BC068239.1 | - | 14 - 15 | 2225 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
