Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800016660.mm.1 |
NM_001001184.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016660.mm.1 |
NM_001001184.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016660.mm.1 |
TC0800002160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016665.mm.1 |
NM_001001184.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016665.mm.1 |
NM_001001184.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016665.mm.1 |
TC0800002160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016656.mm.1 |
NM_001001184.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016656.mm.1 |
NM_001001184.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016656.mm.1 |
TC0800002160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016657.mm.1 |
NM_001001184.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016657.mm.1 |
NM_001001184.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016657.mm.1 |
TC0800002160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016659.mm.1 |
NM_001001184.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016659.mm.1 |
NM_001001184.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016659.mm.1 |
TC0800002160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016658.mm.1 |
NM_001001184.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016658.mm.1 |
NM_001001184.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016658.mm.1 |
TC0800002160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016662.mm.1 |
NM_001001184.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016662.mm.1 |
NM_001001184.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016662.mm.1 |
TC0800002160.mm.1 |
MTA TC |