Assay Details
| Gene Symbol: Enpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177304.3 | NP_796278.1 | 2 - 3 | 467 | 80 | |
| GenBank mRNA | AK038460.1 | - | 2 - 3 | 439 | 80 | |
| AK046797.1 | - | 2 - 3 | 468 | 80 | ||
| AK046881.1 | - | 2 - 3 | 472 | 80 | ||
| AK079352.1 | - | 2 - 3 | 449 | 80 | ||
| AK079642.1 | - | 2 - 3 | 467 | 80 | ||
| BC096376.1 | - | 2 - 3 | 438 | 4237600 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Enpp6
Genomic Map