Assay Details
| Gene Symbol: A1cf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081074.1 | NP_001074543.1 | 3 - 4 | 374 | 109 | |
| GenBank mRNA | AK007970.1 | - | 2 - 3 | 218 | 109 | |
| AY566862.1 | - | 3 - 4 | 371 | 109 | ||
| AY566863.1 | - | 3 - 4 | 371 | 109 | ||
| AY566864.1 | - | 3 - 4 | 371 | 109 | ||
| BC038942.1 | - | 1 - 2 | 74 | 109 | ||
| BC089622.1 | - | 3 - 4 | 322 | 6760455 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

A1cf
Genomic Map