Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500015459.mm.1 |
TC0500001675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015459.mm.1 |
NM_029771.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015459.mm.1 |
NM_029771.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015461.mm.1 |
TC0500001675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015461.mm.1 |
NM_029771.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015461.mm.1 |
NM_029771.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015460.mm.1 |
TC0500001675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015460.mm.1 |
NM_029771.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015460.mm.1 |
NM_029771.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015463.mm.1 |
TC0500001675.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015463.mm.1 |
NM_029771.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015463.mm.1 |
NM_029771.3 |
Refseq |