Assay Details
| Gene Symbol: Taf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081288.1 | NP_001074757.1 | 1 - 2 | 307 | 95 | |
| GenBank mRNA | AK028828.1 | - | 1 - 2 | 54 | 95 | |
| AK147903.1 | - | 1 - 2 | 308 | 95 | ||
| BC147638.1 | - | 1 - 2 | 258 | 9056628 | 95 | |
| BC151079.1 | - | 1 - 2 | 258 | 9087992 | 95 | |
More Information
Set Membership: |
|

Taf2
Genomic Map