Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500011985.mm.1 |
NM_001146180.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011985.mm.1 |
NM_001146180.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011985.mm.1 |
NM_144800.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011985.mm.1 |
NM_144800.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011985.mm.1 |
TC1500001585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500011991.mm.1 |
NM_001146180.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011991.mm.1 |
NM_001146180.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011991.mm.1 |
NM_144800.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011991.mm.1 |
NM_144800.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011991.mm.1 |
TC1500001585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500011989.mm.1 |
NM_001146180.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011989.mm.1 |
NM_001146180.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011989.mm.1 |
NM_144800.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011989.mm.1 |
NM_144800.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011989.mm.1 |
TC1500001585.mm.1 |
MTA TC |