Assay Details
| Gene Symbol: Dmrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015826.5 | NP_056641.2 | 3 - 4 | 970 | 72 | |
| GenBank mRNA | AB221653.1 | - | 3 - 4 | 822 | 72 | |
| AF202778.1 | - | 3 - 4 | 957 | 72 | ||
| AL133300.1 | - | 3 - 4 | 973 | 72 | ||
| AY169786.1 | - | 3 - 4 | 973 | 72 | ||
| AY169787.1 | - | 3 - 4 | 973 | 72 | ||
| BC125474.1 | - | 3 - 4 | 933 | 40129989 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dmrt1
Genomic Map