Assay Details
| Gene Symbol: Ppp1r32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133689.1 | NP_598450.1 | 2 - 3 | 209 | 135 | |
| GenBank mRNA | AK005821.1 | - | 2 - 3 | 241 | 135 | |
| AK016325.1 | - | 2 - 3 | 239 | 135 | ||
| AY032666.1 | - | 2 - 3 | 209 | 135 | ||
| BC118523.1 | - | 2 - 3 | 185 | 40105249 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r32
Genomic Map