Assay Details
| Gene Symbol: Ccdc110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033246.2 | NP_001028418.1 | 2 - 3 | 215 | 108 | |
| GenBank mRNA | AK132742.1 | - | 2 - 3 | 216 | 108 | |
| AK161248.1 | - | 2 - 3 | 203 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc110
Genomic Map