Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800003655.mm.1 |
NM_001033246.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003655.mm.1 |
NM_001033246.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003655.mm.1 |
TC0800000523.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800003657.mm.1 |
NM_001033246.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003657.mm.1 |
NM_001033246.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003657.mm.1 |
TC0800000523.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800003656.mm.1 |
NM_001033246.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003656.mm.1 |
NM_001033246.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800003656.mm.1 |
TC0800000523.mm.1 |
MTA TC |