Assay Details
| Gene Symbol: F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF356627.1 | - | 13 - 14 | 1650 | 78 | |
| AK005546.1 | - | 13 - 14 | 1662 | 78 | ||
| BC019485.1 | - | 13 - 14 | 1639 | 4194672 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

F11
Genomic Map