| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900003344.mm.1 |
NM_013467.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003344.mm.1 |
NM_013467.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003344.mm.1 |
TC1900000335.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003345.mm.1 |
NM_013467.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003345.mm.1 |
NM_013467.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003345.mm.1 |
TC1900000335.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003346.mm.1 |
NM_013467.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003346.mm.1 |
NM_013467.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003346.mm.1 |
TC1900000335.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003347.mm.1 |
NM_013467.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003347.mm.1 |
NM_013467.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003347.mm.1 |
TC1900000335.mm.1 |
MTA TC |